1-Bromo-3-fluoro-2-((methoxymethoxy)methyl)-5-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

55109-500MG 55109-1G 55109-5G 55109-25G
77329-398EA 187.46 USD
77329-398 77353-266 77365-690 77387-614
1-Bromo-3-fluoro-2-((methoxymethoxy)methyl)-5-methylbenzene ≥95%
1-Bromo-3-fluoro-2-((methoxymethoxy)methyl)-5-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFO₂
MW: 263.103 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530355
CAS Number: 2734773-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR