2-Iodo-1,3,5-trimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

55101-500MG 55101-1G 55101-5G 55101-25G
77329-394EA 99.91 USD
77329-394 77342-830 77365-686 77377-516
2-Iodo-1,3,5-trimethoxybenzene ≥97%
2-Iodo-1,3,5-trimethoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₁IO₃
Storage Temperature: Ambient
MDL Number: MFCD00598379
CAS Number: 2510-49-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR