5-(3,5-Dimethylphenyl)-N-methylthiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55099-500MG 55099-1G 55099-5G 55099-25G
77329-392EA 229.69 USD
77329-392 77353-262 77365-684 77387-610
5-(3,5-Dimethylphenyl)-N-methylthiazol-2-amine ≥97%
5-(3,5-Dimethylphenyl)-N-methylthiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂S
MW: 218.318 g/mol
Storage Temperature: Ambient
CAS Number: 1597271-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR