5-(2,3-Dimethylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55066-500MG 55066-1G 55066-5G 55066-25G
77329-388EA 265.74 USD
77329-388 77353-258 77365-680 77387-606
5-(2,3-Dimethylphenyl)thiazol-2-amine ≥97%
5-(2,3-Dimethylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂S
MW: 204.291 g/mol
Storage Temperature: Ambient
CAS Number: 1251361-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR