2-Chloro-3-fluoro-6-methylphenol ≥97%

Supplier: AOBChem USA

Warning

55056-500MG 55056-1G 55056-5G
77337-438EA 414.06 USD
77337-438 77353-256 77377-514
2-Chloro-3-fluoro-6-methylphenol ≥97%
2-Chloro-3-fluoro-6-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD28516576
CAS Number: 1784533-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR