1-(5-(Methylthio)thiophen-2-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

55051-500MG 55051-1G 55051-5G 55051-25G
77329-386EA 136.99 USD
77329-386 77344-540 77365-678 77387-604
1-(5-(Methylthio)thiophen-2-yl)ethanone ≥97%
1-(5-(Methylthio)thiophen-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₈OS₂
MW: 172.268 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02940186
CAS Number: 22102-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR