4'-Bromo-2',5'-dimethyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
55036-500MG
55036-1G
55036-25G
55036-5G
77329-384EA
300.76
USD
77329-384
77353-254
77387-602
77377-510
4'-Bromo-2',5'-dimethyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
4'-Bromo-2',5'-dimethyl-[1,1'-biphenyl]-4-carbaldehyde
Precaución: for Research Use Only
Formula:
C₁₅H₁₃BrO MW: 289.167 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34530348 CAS Number: 2734776-20-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...