4-Bromo-2,4',5-trimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

55035-500MG 55035-1G 55035-5G
77329-382EA 307.97 USD
77329-382 77353-252 77377-508
4-Bromo-2,4',5-trimethyl-1,1'-biphenyl ≥97%
4-Bromo-2,4',5-trimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₅Br
MW: 275.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530347
CAS Number: 864149-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR