4-Bromo-2,2',5-trimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

55034-500MG 55034-1G 55034-5G
77329-380EA 307.97 USD
77329-380 77353-250 77377-506
4-Bromo-2,2',5-trimethyl-1,1'-biphenyl ≥97%
4-Bromo-2,2',5-trimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₅Br
MW: 275.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530346
CAS Number: 2193102-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR