4-Bromo-2,5-dimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

55031-500MG 55031-1G 55031-5G
77329-378EA 332.69 USD
77329-378 77353-248 77377-504
4-Bromo-2,5-dimethyl-1,1'-biphenyl ≥97%
4-Bromo-2,5-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃Br
MW: 261.157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530345
CAS Number: 444683-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR