N-Methyl-5-(m-tolyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55007-500MG 55007-1G 55007-5G 55007-25G
77329-376EA 204.97 USD
77329-376 77353-246 77365-676 77387-600
N-Methyl-5-(m-tolyl)thiazol-2-amine ≥97%
N-Methyl-5-(m-tolyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂S
MW: 204.291 g/mol
Storage Temperature: Ambient
CAS Number: 1602610-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR