N-Methyl-5-(p-tolyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55006-500MG 55006-1G 55006-5G 55006-25G
77329-374EA 204.97 USD
77329-374 77353-244 77365-674 77387-598
N-Methyl-5-(p-tolyl)thiazol-2-amine ≥97%
N-Methyl-5-(p-tolyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂S
MW: 204.291 g/mol
Storage Temperature: Ambient
CAS Number: 1600075-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR