Ethyl 2,3-dibromo-5-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

54998-500MG 54998-1G 54998-5G 54998-25G
77329-372EA 90.64 USD
77329-372 77342-396 77365-672 77377-502
Ethyl 2,3-dibromo-5-methylbenzoate ≥95%
Ethyl 2,3-dibromo-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀Br₂O₂
MW: 321.993 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24107272
CAS Number: 1542608-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR