N-Methyl-5-phenylthiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

54971-500MG 54971-1G 54971-5G 54971-25G
77329-366EA 256.47 USD
77329-366 77353-240 77365-664 77387-594
N-Methyl-5-phenylthiazol-2-amine ≥97%
N-Methyl-5-phenylthiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂S
MW: 190.265 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17258258
CAS Number: 1344198-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR