5-(M-tolyl)thiazol-2-amine

Supplier: AOBChem USA

Warning

54968-500MG 54968-1G 54968-5G
77337-758EA 440.84 USD
77337-758 77353-238 77377-492
5-(M-tolyl)thiazol-2-amine
5-(M-tolyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂S
MW: 190.265 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12824102
CAS Number: 1178168-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR