(4-Bromo-2-fluoro-5-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

54916-500MG 54916-1G 54916-5G
77338-496EA 491.31 USD
77338-496 77353-232 77377-486
(4-Bromo-2-fluoro-5-methylphenyl)(methyl)sulfane ≥97%
(4-Bromo-2-fluoro-5-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈BrFS
Storage Temperature: Ambient
MDL Number: MFCD28403624
CAS Number: 1011460-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR