(4-Chloro-5-fluoro-2-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

54905-500MG 54905-1G 54905-5G 54905-25G
77329-354EA 189.52 USD
77329-354 77353-230 77365-652 77387-582
(4-Chloro-5-fluoro-2-methylphenyl)(methyl)sulfane ≥97%
(4-Chloro-5-fluoro-2-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈ClFS
MW: 190.666 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28752242
CAS Number: 1807037-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR