4-Chloro-5-fluoro-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

54898-500MG 54898-1G 54898-5G
77329-352EA 319.3 USD
77329-352 77353-228 77377-484
4-Chloro-5-fluoro-2-methylbenzaldehyde ≥97%
4-Chloro-5-fluoro-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD28553442
CAS Number: 1780682-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR