Methyl 3-fluoro-5-methoxy-4-methylbenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
54891-500MG
54891-1G
54891-5G
77339-760EA
631.39
USD
77339-760
77353-226
77377-482
Methyl 3-fluoro-5-methoxy-4-methylbenzoate ≥97%
Methyl 3-fluoro-5-methoxy-4-methylbenzoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₁FO₃ MW: 198.191 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34757410 CAS Number: 1818898-20-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...