2,2,2-Trifluoro-1-(3-methoxy-2,4-dimethylphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

54879-500MG 54879-1G 54879-5G
77340-218EA 839.45 USD
77340-218 77353-224 77377-480
2,2,2-Trifluoro-1-(3-methoxy-2,4-dimethylphenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(3-methoxy-2,4-dimethylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₁H₁₁F₃O₂
MW: 232.199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757405
CAS Number: 2901101-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR