2,2,2-Trifluoro-1-(3-fluoro-2,4-dimethylphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

54874-500MG 54874-1G 54874-5G
77337-706EA 437.75 USD
77337-706 77353-222 77377-478
2,2,2-Trifluoro-1-(3-fluoro-2,4-dimethylphenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(3-fluoro-2,4-dimethylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₈F₄O
MW: 220.164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757401
CAS Number: 2901085-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR