1-(4-Fluoro-3,5-dimethylphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

54871-500MG 54871-1G 54871-5G 54871-25G
77329-348EA 161.71 USD
77329-348 77345-886 77365-648 77387-578
1-(4-Fluoro-3,5-dimethylphenyl)ethanol ≥95%
1-(4-Fluoro-3,5-dimethylphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₃FO
MW: 168.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21187170
CAS Number: 1443344-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR