2-(4-Fluoro-3,5-dimethylphenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

54870-500MG 54870-1G 54870-5G 54870-25G
77329-346EA 204.97 USD
77329-346 77353-220 77365-646 77387-576
2-(4-Fluoro-3,5-dimethylphenyl)propan-2-ol ≥97%
2-(4-Fluoro-3,5-dimethylphenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₁₁H₁₅FO
MW: 182.235 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757399
CAS Number: 2807465-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR