2-Chloro-6-fluoro-4-methylphenol ≥97%

Supplier: AOBChem USA

Warning

54864-500MG 54864-1G 54864-5G
77329-344EA 319.3 USD
77329-344 77353-216 77377-474
2-Chloro-6-fluoro-4-methylphenol ≥97%
2-Chloro-6-fluoro-4-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD22414680
CAS Number: 1805042-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR