3-Ethyl-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

54858-500MG 54858-1G 54858-5G
77339-166EA 556.2 USD
77339-166 77353-214 77377-472
3-Ethyl-5-methylbenzoic acid ≥97%
3-Ethyl-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12547781
CAS Number: 1208076-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR