(2-Bromo-6-fluoro-4-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

54844-1G 54844-5G 54844-25G
77340-736EA 60.77 USD
77340-736 77365-640 77377-468
(2-Bromo-6-fluoro-4-methylphenyl)(methyl)sulfane ≥97%
(2-Bromo-6-fluoro-4-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈BrFS
Storage Temperature: Ambient
MDL Number: MFCD34530319
CAS Number: 2706448-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR