1-(2-Bromo-6-fluoro-4-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

54843-1G 54843-5G 54843-25G
77341-388EA 83.43 USD
77341-388 77365-638 77377-466
1-(2-Bromo-6-fluoro-4-methylphenyl)ethanone ≥95%
1-(2-Bromo-6-fluoro-4-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₈BrFO
MW: 231.062 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28579982
CAS Number: 1935407-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR