5-Ethoxy-2,4-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

54807-500MG 54807-1G
77340-496EA 1282.35 USD
77340-496 77353-210
5-Ethoxy-2,4-dimethylbenzoic acid ≥97%
5-Ethoxy-2,4-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD34757387
CAS Number: 2091216-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR