2,2-Dichloro-1-(2-chloro-5-(trifluoromethyl)phenyl)ethanol ≥95%
Supplier: AOBChem USA
|
Warning
|
54796-500MG
54796-1G
54796-5G
54796-25G
77329-340EA
99.91
USD
77329-340
77342-828
77365-632
77377-456
2,2-Dichloro-1-(2-chloro-5-(trifluoromethyl)phenyl)ethanol ≥95%
2,2-Dichloro-1-(2-chloro-5-(trifluoromethyl)phenyl)ethanol
Precaución: for Research Use Only
Formula:
C₉H₆Cl₃F₃O MW: 293.498 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34530310 CAS Number: 2734773-64-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...