Methyl 2,5-dichloro-4-methoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

54786-500MG 54786-1G 54786-5G 54786-25G
77329-336EA 257.5 USD
77329-336 77353-204 77365-630 77387-572
Methyl 2,5-dichloro-4-methoxybenzoate ≥95%
Methyl 2,5-dichloro-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD22988334
CAS Number: 1228376-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR