(2,5-Dichloro-4-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

54782-500MG 54782-1G 54782-5G
77335-988EA 372.86 USD
77335-988 77353-202 77377-452
(2,5-Dichloro-4-methoxyphenyl)(methyl)sulfane ≥97%
(2,5-Dichloro-4-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈Cl₂OS
MW: 223.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530307
CAS Number: 2709762-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR