Ethyl 2,5-dichloro-4-ethoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

54778-500MG 54778-1G 54778-5G
77336-912EA 399.64 USD
77336-912 77353-198 77377-448
Ethyl 2,5-dichloro-4-ethoxybenzoate ≥97%
Ethyl 2,5-dichloro-4-ethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34757386
CAS Number: 2387021-66-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR