2,5-Dichloro-4-ethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

54776-500MG 54776-1G 54776-5G
77339-294EA 572.68 USD
77339-294 77353-194 77377-444
2,5-Dichloro-4-ethoxybenzoic acid ≥97%
2,5-Dichloro-4-ethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD24248175
CAS Number: 1557678-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR