(2,5-Dichloro-4-ethoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

54774-500MG 54774-1G 54774-5G
77337-436EA 414.06 USD
77337-436 77353-192 77377-442
(2,5-Dichloro-4-ethoxyphenyl)boronic acid ≥97%
(2,5-Dichloro-4-ethoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BCl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD24248207
CAS Number: 2246664-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR