1-Bromo-2,5-dichloro-4-(difluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

54770-500MG 54770-1G 54770-5G 54770-25G
77329-332EA 218.36 USD
77329-332 77353-188 77365-626 77387-570
1-Bromo-2,5-dichloro-4-(difluoromethoxy)benzene ≥97%
1-Bromo-2,5-dichloro-4-(difluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₃BrCl₂F₂O
MW: 291.905 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32703116
CAS Number: 2168997-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR