2,5-Dichloro-4-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

54767-500MG 54767-1G 54767-5G
77339-436EA 590.19 USD
77339-436 77353-186 77377-436
2,5-Dichloro-4-(methylthio)phenol ≥97%
2,5-Dichloro-4-(methylthio)phenol

Precaución: for Research Use Only

Formula: C₇H₆Cl₂OS
MW: 209.093 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24714448
CAS Number: 21923-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR