4-((4-Fluorobenzyl)oxy)-2-methoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

54754-500MG 54754-1G 54754-5G 54754-25G
77329-330EA 150.38 USD
77329-330 77345-354 77365-624 77387-568
4-((4-Fluorobenzyl)oxy)-2-methoxybenzaldehyde ≥95%
4-((4-Fluorobenzyl)oxy)-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₃
MW: 260.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530305
CAS Number: 2734773-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR