4-Bromo-3,5-dichloro-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

54751-500MG 54751-1G 54751-25G 54751-5G
77329-326EA 284.28 USD
77329-326 77353-184 77387-566 77377-432
4-Bromo-3,5-dichloro-1,1'-biphenyl ≥95%
4-Bromo-3,5-dichloro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₇BrCl₂
MW: 301.994 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530303
CAS Number: 2445503-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR