5-Iodo-2-methoxy-1,3-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

54748-1G 54748-5G 54748-25G
77341-526EA 91.67 USD
77341-526 77365-618 77377-428
5-Iodo-2-methoxy-1,3-dimethylbenzene ≥97%
5-Iodo-2-methoxy-1,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁IO
Storage Temperature: Ambient
MDL Number: MFCD07779018
CAS Number: 55215-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR