3,5-Difluoro-4-methoxy-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

54743-500MG 54743-1G 54743-5G 54743-25G
77329-324EA 281.19 USD
77329-324 77353-182 77365-614 77387-564
3,5-Difluoro-4-methoxy-1,1'-biphenyl ≥95%
3,5-Difluoro-4-methoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₂O
MW: 220.215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530301
CAS Number: 2734775-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR