3',5'-Difluoro-4'-methoxy-2-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

54741-500MG 54741-1G 54741-5G 54741-25G
77329-322EA 225.57 USD
77329-322 77353-180 77365-612 77387-562
3',5'-Difluoro-4'-methoxy-2-methyl-1,1'-biphenyl ≥97%
3',5'-Difluoro-4'-methoxy-2-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₂O
MW: 234.241 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530300
CAS Number: 2734779-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR