3,5-Difluoro-4-methoxy-4'-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

54740-500MG 54740-1G 54740-5G 54740-25G
77329-320EA 225.57 USD
77329-320 77353-178 77365-610 77387-560
3,5-Difluoro-4-methoxy-4'-methyl-1,1'-biphenyl ≥97%
3,5-Difluoro-4-methoxy-4'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₂O
MW: 234.241 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530299
CAS Number: 2734772-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR