4-Chloro-2-fluoro-3-methyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

54702-500MG 54702-1G 54702-5G
77338-872EA 527.36 USD
77338-872 77353-174 77377-422
4-Chloro-2-fluoro-3-methyl-1,1'-biphenyl ≥95%
4-Chloro-2-fluoro-3-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀ClF
MW: 220.67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530297
CAS Number: 2734772-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR