4'-Chloro-2'-fluoro-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

54700-500MG 54700-1G 54700-5G 54700-25G
77329-316EA 200.85 USD
77329-316 77353-172 77365-606 77387-556
4'-Chloro-2'-fluoro-[1,1'-biphenyl]-4-carbaldehyde ≥97%
4'-Chloro-2'-fluoro-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₈ClFO
MW: 234.653 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23852995
CAS Number: 1432737-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR