2-(Benzyloxy)-5-bromo-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

54677-500MG 54677-1G 54677-5G 54677-25G
77329-314EA 141.11 USD
77329-314 77344-836 77365-604 77387-554
2-(Benzyloxy)-5-bromo-3-methoxybenzaldehyde ≥97%
2-(Benzyloxy)-5-bromo-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃BrO₃
MW: 321.166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09721102
CAS Number: 155351-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR