2,5-Dichloro-3-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

54670-500MG 54670-1G 54670-5G
77339-780EA 638.6 USD
77339-780 77353-170 77377-420
2,5-Dichloro-3-iodobenzaldehyde ≥97%
2,5-Dichloro-3-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃Cl₂IO
MW: 300.909 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32705776
CAS Number: 2092184-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR