2-Chloro-1-fluoro-5-iodo-3-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

54651-500MG 54651-1G 54651-5G 54651-25G
77329-310EA 90.64 USD
77329-310 77342-394 77365-598 77377-414
2-Chloro-1-fluoro-5-iodo-3-methoxybenzene ≥95%
2-Chloro-1-fluoro-5-iodo-3-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅ClFIO
MW: 286.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757359
CAS Number: 2387014-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR