(2-Propoxy-4-(trifluoromethyl)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

54645-500MG 54645-1G 54645-5G
77338-630EA 504.7 USD
77338-630 77353-166 77377-410
(2-Propoxy-4-(trifluoromethyl)phenyl)methanol ≥97%
(2-Propoxy-4-(trifluoromethyl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₁₃F₃O₂
MW: 234.215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564273
CAS Number: 2734773-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR