Methyl(2-propoxy-4-(trifluoromethyl)phenyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

54643-500MG 54643-1G 54643-5G
77339-304EA 574.74 USD
77339-304 77353-162 77377-406
Methyl(2-propoxy-4-(trifluoromethyl)phenyl)sulfane ≥97%
Methyl(2-propoxy-4-(trifluoromethyl)phenyl)sulfane

Precaución: for Research Use Only

Formula: C₁₁H₁₃F₃OS
MW: 250.281 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757357
CAS Number: 2807450-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR