1-Iodo-2-propoxy-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

54642-500MG 54642-1G 54642-5G
77329-308EA 338.87 USD
77329-308 77353-160 77377-404
1-Iodo-2-propoxy-4-(trifluoromethyl)benzene ≥97%
1-Iodo-2-propoxy-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₃IO
MW: 330.085 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530290
CAS Number: 2384902-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR